Ethyl 2-(2-chloro-4-methoxyphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

147492-5G 147492-1G 147492-500MG
77631-322EA 1516.16 USD
77631-322 77636-660 77652-354
Ethyl 2-(2-chloro-4-methoxyphenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-chloro-4-methoxyphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 242.656 g/mol
MW: C₁₁H₁₁ClO₄
Storage Temperature: Ambient
MDL Number: MFCD16061592
CAS Number: 1249917-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR