Ethyl 2-(3-chloro-4-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

147491-5G 147491-25G 147491-1G 147491-500MG
77631-320EA 1484.23 USD
77631-320 77635-452 77640-672 77644-642
Ethyl 2-(3-chloro-4-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-chloro-4-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 280.628 g/mol
MW: C₁₁H₈ClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34839401
CAS Number: 2921750-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR