Ethyl 2-(3-(methoxymethoxy)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

147485-5G 147485-25G 147485-1G 147485-500MG
77631-318EA 1667.57 USD
77631-318 77635-450 77636-658 77648-688
Ethyl 2-(3-(methoxymethoxy)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-(methoxymethoxy)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 238.237 g/mol
MW: C₁₂H₁₄O₅
Storage Temperature: Ambient
MDL Number: MFCD34839395
CAS Number: 2921899-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR