4-(2-Bromo-5-chloropyridin-4-yl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

147105-5G 147105-1G 147105-25G 147105-500MG
77631-316EA 1667.57 USD
77631-316 77636-656 77653-588 77644-640
4-(2-Bromo-5-chloropyridin-4-yl)tetrahydro-2H-pyran-4-ol ≥95%
4-(2-Bromo-5-chloropyridin-4-yl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: C₁₀H₁₁BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD34839232
CAS Number: 2921719-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR