2,2'-Bithiophene-5-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-([2,2'-Bithiophen]-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

14700-5G 14700-25G
77634-042EA 96 USD
77634-042 77657-470
2,2'-Bithiophene-5-boronic acid pinacol ester ≥97%
2,2'-Bithiophene-5-boronic acid pinacol ester

Precaución: For Research Use Only

Formula: C₁₄H₁₇BO₂S₂
Storage Temperature: Ambient
MDL Number: MFCD04039974
CAS Number: 479719-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR