1-(5-Bromo-2-chloropyridin-4-yl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

146753-5G 146753-25G 146753-1G 146753-500MG
77631-314EA 1667.57 USD
77631-314 77653-586 77640-668 77648-686
1-(5-Bromo-2-chloropyridin-4-yl)cyclohexanol ≥97%
1-(5-Bromo-2-chloropyridin-4-yl)cyclohexanol

Precaución: For Research Use Only

Formula: 290.584 g/mol
MW: C₁₁H₁₃BrClNO
Storage Temperature: Ambient
MDL Number: MFCD34838950
CAS Number: 2921879-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR