Ethyl 2-(2,6-dichloropyridin-3-yl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

146726-5G 146726-25G 146726-1G 146726-500MG
77631-312EA 1667.57 USD
77631-312 77635-448 77640-666 77648-684
Ethyl 2-(2,6-dichloropyridin-3-yl)-2-oxoacetate ≥95%
Ethyl 2-(2,6-dichloropyridin-3-yl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₉H₇Cl₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD12032193
CAS Number: 1260778-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR