3-Bromo-5-(trimethylsilyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

146675-5G 146675-1G 146675-500MG
77653-584EA 1898 USD
77653-584 77644-636 77644-638
3-Bromo-5-(trimethylsilyl)pyridine ≥97%
3-Bromo-5-(trimethylsilyl)pyridine

Precaución: For Research Use Only

Formula: C₈H₁₂BrNSi
Storage Temperature: Ambient
MDL Number: MFCD12028716
CAS Number: 144056-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR