Methyl 3-fluoro-5-methoxyisonicotinate ≥97%

Supplier: AOBChem USA

Warning

146635-5G 146635-1G 146635-500MG
77631-310EA 1667.57 USD
77631-310 77640-664 77644-634
Methyl 3-fluoro-5-methoxyisonicotinate ≥97%
Methyl 3-fluoro-5-methoxyisonicotinate

Precaución: For Research Use Only

Formula: C₈H₈FNO₃
Storage Temperature: Ambient
MDL Number: MFCD18258151
CAS Number: 1256787-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR