3-Fluoro-5-methoxyisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

146634-5G 146634-1G 146634-500MG
77657-468EA 2579 USD
77657-468 77640-662 77644-632
3-Fluoro-5-methoxyisonicotinic acid ≥97%
3-Fluoro-5-methoxyisonicotinic acid

Precaución: For Research Use Only

Formula: C₇H₆FNO₃
Storage Temperature: Ambient
MDL Number: MFCD18257427
CAS Number: 1256810-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR