6-Chloropiperonal ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-1,3-benzodioxole-5-carboxaldehyde

146132-5G 146132-1G 146132-25G 146132-500MG
77634-040EA 326 USD
77634-040 77636-654 77653-580 77644-630
6-Chloropiperonal ≥97%
6-Chloropiperonal

Precaución: For Research Use Only

Formula: C₈H₅ClO₃
MW: 184.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022953
CAS Number: 15952-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR