(5-Bromo-2-(trifluoromethyl)phenyl)(isopropyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

145900-5G 145900-25G 145900-1G 145900-500MG
77631-308EA 1667.57 USD
77631-308 77653-578 77640-660 77648-682
(5-Bromo-2-(trifluoromethyl)phenyl)(isopropyl)sulfane ≥97%
(5-Bromo-2-(trifluoromethyl)phenyl)(isopropyl)sulfane

Precaución: For Research Use Only

Formula: 299.151 g/mol
MW: C₁₀H₁₀BrF₃S
Storage Temperature: Ambient
MDL Number: MFCD34838313
CAS Number: 2921838-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR