2-(Benzyloxy)-1,3-dibromo-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

14565-5G 14565-1G 14565-25G 14565-500MG
77634-038EA 297 USD
77634-038 77636-652 77657-466 77648-680
2-(Benzyloxy)-1,3-dibromo-5-methylbenzene ≥97%
2-(Benzyloxy)-1,3-dibromo-5-methylbenzene

Precaución: For Research Use Only

Formula: C₁₄H₁₂Br₂O
MW: 356.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00187381
CAS Number: 84379-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR