2-Bromo-4-methyl-1-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

14562-5G 14562-25G 14562-1G 14562-500MG
77631-658EA 1050 USD
77631-658 77657-464 77640-658 77648-678
2-Bromo-4-methyl-1-propoxybenzene ≥95%
2-Bromo-4-methyl-1-propoxybenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11156047
CAS Number: 99187-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR