(E)-1-(2,5-difluorophenyl)but-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

145314-5G 145314-1G 145314-500MG
77631-304EA 1484.23 USD
77631-304 77640-656 77648-676
(E)-1-(2,5-difluorophenyl)but-2-en-1-ol ≥95%
(E)-1-(2,5-difluorophenyl)but-2-en-1-ol

Precaución: For Research Use Only

Formula: 184.183 g/mol
MW: C₁₀H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD30678741
CAS Number: 2276728-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR