2-(2-Bromo-5-methylphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

14492-5G 14492-1G 14492-25G 14492-500MG
77634-036EA 536 USD
77634-036 77636-646 77657-460 77644-622
2-(2-Bromo-5-methylphenyl)-1,3-dioxolane ≥95%
2-(2-Bromo-5-methylphenyl)-1,3-dioxolane

Precaución: For Research Use Only

Formula: 243.097 g/mol
MW: C₁₀H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD31619360
CAS Number: 1381966-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR