5-Bromo-2-(piperidin-1-yl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

14466-5G 14466-1G 14466-25G 14466-500MG
77634-034EA 424 USD
77634-034 77636-644 77653-570 77648-666
5-Bromo-2-(piperidin-1-yl)benzonitrile ≥95%
5-Bromo-2-(piperidin-1-yl)benzonitrile

Precaución: For Research Use Only

Formula: C₁₂H₁₃BrN₂
Storage Temperature: Ambient
MDL Number: MFCD11037627
CAS Number: 876918-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR