(3-(Benzyloxy)pyridin-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

144385-5G 144385-1G 144385-500MG
77653-568EA 1854 USD
77653-568 77640-650 77648-664
(3-(Benzyloxy)pyridin-2-yl)methanol ≥97%
(3-(Benzyloxy)pyridin-2-yl)methanol

Precaución: For Research Use Only

Formula: 215.248 g/mol
MW: C₁₃H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD16251531
CAS Number: 6059-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR