(3,4-Dihydro-2H-benzo[b][1,4]dioxepin-6-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

14421-5G 14421-1G 14421-25G 14421-500MG
77634-030EA 404 USD
77634-030 77636-642 77657-458 77644-620
(3,4-Dihydro-2H-benzo[b][1,4]dioxepin-6-yl)boronic acid ≥95%
(3,4-Dihydro-2H-benzo[b][1,4]dioxepin-6-yl)boronic acid

Precaución: For Research Use Only

Formula: C₉H₁₁BO₄
MW: 193.9922 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06801681
CAS Number: 1107064-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR