1-Bromo-2-iodo-5-methoxy-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

144148-5G 144148-1G 144148-25G 144148-500MG
77631-298EA 1823.1 USD
77631-298 77636-640 77653-564 77644-618
1-Bromo-2-iodo-5-methoxy-4-(trifluoromethyl)benzene ≥97%
1-Bromo-2-iodo-5-methoxy-4-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 380.928 g/mol
MW: C₈H₅BrF₃IO
Storage Temperature: Ambient
MDL Number: MFCD34837095
CAS Number: 2385224-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR