5-Methylnicotinohydrazide ≥97%

Supplier: AOBChem USA

Warning

14374-5G 14374-1G 14374-25G 14374-500MG
77631-654EA 876 USD
77631-654 77636-638 77653-562 77648-662
5-Methylnicotinohydrazide ≥97%
5-Methylnicotinohydrazide

Precaución: For Research Use Only

Formula: C₇H₉N₃O
Storage Temperature: Ambient
MDL Number: MFCD00831081
CAS Number: 808127-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR