1-(6-Chloro-2-fluoropyridin-3-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

142654-5G 142654-25G 142654-1G 142654-500MG
77631-648EA 1024 USD
77631-648 77653-558 77640-644 77644-616
1-(6-Chloro-2-fluoropyridin-3-yl)ethanone ≥95%
1-(6-Chloro-2-fluoropyridin-3-yl)ethanone

Precaución: For Research Use Only

Formula: C₇H₅ClFNO
Storage Temperature: Ambient
MDL Number: MFCD18250252
CAS Number: 1260663-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR