3'-Chloro-5'-methoxyacetophenone ≥95%
Supplier: AOBChem USA
|
Warning
|
142641-5G
142641-25G
142641-1G
142641-500MG
77631-646EA
882
USD
77631-646
77653-556
77640-642
77648-654
3'-Chloro-5'-methoxyacetophenone ≥95%
3'-Chloro-5'-methoxyacetophenone
Precaución: For Research Use Only
Formula:
C₉H₉ClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD22544395 CAS Number: 214760-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...