3-Isopropoxy-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

14250-5G 14250-1G 14250-25G 14250-500MG
77631-644EA 825 USD
77631-644 77636-634 77657-452 77648-652
3-Isopropoxy-4-methylbenzaldehyde ≥97%
3-Isopropoxy-4-methylbenzaldehyde

Precaución: For Research Use Only

Formula: 178.228 g/mol
MW: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD24685689
CAS Number: 532965-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR