3-Bromo-5-(trifluoromethoxy)toluene ≥95%

Supplier: AOBChem USA

Warning

142497-5G 142497-25G
77634-024EA 160 USD
77634-024 77653-554
3-Bromo-5-(trifluoromethoxy)toluene ≥95%
3-Bromo-5-(trifluoromethoxy)toluene

Precaución: For Research Use Only

Formula: C₈H₆BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD07368791
CAS Number: 887266-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR