1-[2,5-Bis(trifluoromethyl)phenyl]ethanol ≥97%

Supplier: AOBChem USA

Warning

142404-5G 142404-25G
77634-022EA 108 USD
77634-022 77653-552
1-[2,5-Bis(trifluoromethyl)phenyl]ethanol ≥97%
1-[2,5-Bis(trifluoromethyl)phenyl]ethanol

Precaución: For Research Use Only

Formula: C₁₀H₈F₆O
MW: 258.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10686545
CAS Number: 364-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR