6-Chloro-2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

142313-1G 142313-500MG 142313-5G
77636-632EA 657 USD
77636-632 77652-350 77653-550
6-Chloro-2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylic acid ≥97%
6-Chloro-2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylic acid

Precaución: For Research Use Only

Formula: 214.602 g/mol
MW: C₉H₇ClO₄
Storage Temperature: Ambient
MDL Number: MFCD24497993
CAS Number: 66411-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR