1-(5-Chlorobenzo[d][1,3]dioxol-4-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

142311-5G 142311-1G 142311-25G 142311-500MG
77631-642EA 1206 USD
77631-642 77636-630 77657-448 77648-648
1-(5-Chlorobenzo[d][1,3]dioxol-4-yl)ethanone ≥95%
1-(5-Chlorobenzo[d][1,3]dioxol-4-yl)ethanone

Precaución: For Research Use Only

Formula: 198.603 g/mol
MW: C₉H₇ClO₃
Storage Temperature: Ambient
MDL Number: MFCD32706366
CAS Number: 1554250-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR