5-Chloro-4-(methylthio)benzo[d][1,3]dioxole ≥95%
Supplier: AOBChem USA
|
Warning
|
142308-5G
142308-25G
142308-1G
142308-500MG
77631-294EA
1484.23
USD
77631-294
77635-444
77636-628
77648-646
5-Chloro-4-(methylthio)benzo[d][1,3]dioxole ≥95%
5-Chloro-4-(methylthio)benzo[d][1,3]dioxole
Precaución: For Research Use Only
Formula:
202.658 g/mol MW: C₈H₇ClO₂S Storage Temperature: Ambient |
MDL Number:
MFCD34835715 CAS Number: 2921875-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...