2-Chloro-5-(trifluoromethoxy)aniline ≥97%

Supplier: AOBChem USA

Warning

14229-5G 14229-1G 14229-25G
77634-020EA 205 USD
77634-020 77636-626 77653-548
2-Chloro-5-(trifluoromethoxy)aniline ≥97%
2-Chloro-5-(trifluoromethoxy)aniline

Precaución: For Research Use Only

Formula: C₇H₅ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD16658882
CAS Number: 331-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR