1-Bromo-3-fluoro-5-methoxy-2-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

142059-5G 142059-25G 142059-1G 142059-500MG
77631-292EA 1484.23 USD
77631-292 77657-446 77640-636 77648-644
1-Bromo-3-fluoro-5-methoxy-2-(methoxymethoxy)benzene ≥95%
1-Bromo-3-fluoro-5-methoxy-2-(methoxymethoxy)benzene

Precaución: For Research Use Only

Formula: 265.076 g/mol
MW: C₉H₁₀BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD34835550
CAS Number: 2921730-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR