4,5-Dimethoxyphthalaldehyde ≥97%

Supplier: AOBChem USA

Warning

14173-1G 14173-5G 14173-500MG
77636-624EA 766 USD
77636-624 77653-544 77648-642
4,5-Dimethoxyphthalaldehyde ≥97%
4,5-Dimethoxyphthalaldehyde

Precaución: For Research Use Only

Formula: C₁₀H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00185592
CAS Number: 43073-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR