1-(5-Phenoxypyridin-3-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

141456-5G 141456-25G 141456-1G 141456-500MG
77631-290EA 1484.23 USD
77631-290 77653-542 77640-634 77648-638
1-(5-Phenoxypyridin-3-yl)ethanone ≥95%
1-(5-Phenoxypyridin-3-yl)ethanone

Precaución: For Research Use Only

Formula: 213.232 g/mol
MW: C₁₃H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD34835197
CAS Number: 2011201-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR