Methyl(4-(tert-pentyl)phenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

140755-5G 140755-25G 140755-1G 140755-500MG
77631-288EA 1588.26 USD
77631-288 77635-442 77640-630 77648-632
Methyl(4-(tert-pentyl)phenyl)sulfane ≥95%
Methyl(4-(tert-pentyl)phenyl)sulfane

Precaución: For Research Use Only

Formula: C₁₂H₁₈S
Storage Temperature: Ambient
MDL Number: MFCD34834786
CAS Number: 2921852-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR