6-Bromo-2-fluoro-3-isopropylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

140738-5G 140738-25G 140738-1G 140738-500MG
77631-286EA 1806.62 USD
77631-286 77635-440 77640-626 77648-628
6-Bromo-2-fluoro-3-isopropylbenzaldehyde ≥95%
6-Bromo-2-fluoro-3-isopropylbenzaldehyde

Precaución: For Research Use Only

Formula: C₁₀H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD34834777
CAS Number: 2921687-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR