3-Fluoro-4-isopropylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

140728-1G 140728-5G 140728-500MG
77634-018EA 957 USD
77634-018 77653-536 77644-608
3-Fluoro-4-isopropylbenzaldehyde ≥95%
3-Fluoro-4-isopropylbenzaldehyde

Precaución: For Research Use Only

Formula: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD24107875
CAS Number: 1289156-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR