4-Bromo-3-chloro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

14046-5G 14046-1G 14046-25G 14046-500MG
77634-016EA 400 USD
77634-016 77636-622 77653-534 77648-626
4-Bromo-3-chloro-2-methylbenzaldehyde ≥97%
4-Bromo-3-chloro-2-methylbenzaldehyde

Precaución: For Research Use Only

Formula: 233.49 g/mol
MW: C₈H₆BrClO
Storage Temperature: Ambient
MDL Number: MFCD09835095
CAS Number: 1224604-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR