2-Ethoxy-1-iodo-4-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

140374-5G 140374-1G 140374-25G 140374-500MG
77631-284EA 1806.62 USD
77631-284 77636-620 77657-444 77644-606
2-Ethoxy-1-iodo-4-methoxybenzene ≥95%
2-Ethoxy-1-iodo-4-methoxybenzene

Precaución: For Research Use Only

Formula: 278.087 g/mol
MW: C₉H₁₁IO₂
Storage Temperature: Ambient
MDL Number: MFCD34834510
CAS Number: 35287-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR