(2-Ethoxy-4-methoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

140373-5G 140373-25G 140373-1G 140373-500MG
77631-638EA 1441 USD
77631-638 77653-532 77640-624 77644-604
(2-Ethoxy-4-methoxyphenyl)(methyl)sulfane ≥95%
(2-Ethoxy-4-methoxyphenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: C₁₀H₁₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD34834509
CAS Number: 2921887-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR