2,2-Dimethyl-1-(naphthalen-2-yl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140289-5G 140289-1G 140289-25G 140289-500MG
77631-282EA 1806.62 USD
77631-282 77636-618 77653-530 77648-624
2,2-Dimethyl-1-(naphthalen-2-yl)propan-1-ol ≥95%
2,2-Dimethyl-1-(naphthalen-2-yl)propan-1-ol

Precaución: For Research Use Only

Formula: 214.303 g/mol
MW: C₁₅H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD16847356
CAS Number: 100008-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR