2,2-Dimethyl-1-(naphthalen-1-yl)propan-1-ol ≥97%

Supplier: AOBChem USA

Warning

140288-5G 140288-1G 140288-25G 140288-500MG
77631-280EA 1806.62 USD
77631-280 77636-616 77653-528 77648-622
2,2-Dimethyl-1-(naphthalen-1-yl)propan-1-ol ≥97%
2,2-Dimethyl-1-(naphthalen-1-yl)propan-1-ol

Precaución: For Research Use Only

Formula: 214.303 g/mol
MW: C₁₅H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD16847326
CAS Number: 112825-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR