1-(4-Chloro-2-methoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140259-5G 140259-1G 140259-25G 140259-500MG
77631-636EA 1206 USD
77631-636 77636-614 77653-526 77648-620
1-(4-Chloro-2-methoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-Chloro-2-methoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For Research Use Only

Formula: 228.715 g/mol
MW: C₁₂H₁₇ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34834432
CAS Number: 1599381-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR