1-(4-Methoxy-2-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140233-5G 140233-25G 140233-1G 140233-500MG
77631-634EA 1206 USD
77631-634 77657-442 77640-622 77644-602
1-(4-Methoxy-2-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-Methoxy-2-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For Research Use Only

Formula: C₁₃H₂₀O₂
MW: 208.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787103
CAS Number: 1308026-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR