1-(2,4-Dimethylphenyl)-2,2-dimethylpropan-1-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
140211-5G
140211-1G
140211-25G
140211-500MG
77631-278EA
1484.23
USD
77631-278
77636-610
77657-438
77644-600
1-(2,4-Dimethylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2,4-Dimethylphenyl)-2,2-dimethylpropan-1-ol
Precaución: For Research Use Only
Formula:
C₁₃H₂₀O Storage Temperature: Ambient |
MDL Number:
MFCD12787082 CAS Number: 854169-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...