1-(4-Ethoxyphenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

140193-5G 140193-1G 140193-25G 140193-500MG
77631-630EA 904 USD
77631-630 77636-608 77657-436 77648-616
1-(4-Ethoxyphenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(4-Ethoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For Research Use Only

Formula: 208.297 g/mol
MW: C₁₃H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD12786985
CAS Number: 1225688-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR