1-(3-(Benzyloxy)phenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140181-5G 140181-1G 140181-25G 140181-500MG
77631-628EA 1206 USD
77631-628 77636-606 77657-434 77648-614
1-(3-(Benzyloxy)phenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-(Benzyloxy)phenyl)-2,2-dimethylpropan-1-ol

Precaución: For Research Use Only

Formula: C₁₈H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34834407
CAS Number: 1882016-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR