3-Fluoro-4-(trimethylsilyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

13996-5G 13996-25G 13996-1G 13996-500MG
77631-626EA 841 USD
77631-626 77657-432 77640-618 77644-596
3-Fluoro-4-(trimethylsilyl)pyridine ≥95%
3-Fluoro-4-(trimethylsilyl)pyridine

Precaución: For Research Use Only

Formula: 169.271 g/mol
MW: C₈H₁₂FNSi
Storage Temperature: Ambient
MDL Number: MFCD18803377
CAS Number: 77332-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR