2-Chloro-3-fluoro-4-(trimethylsilyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

13995-5G 13995-25G 13995-1G 13995-500MG
77631-276EA 1583.11 USD
77631-276 77653-522 77640-616 77644-594
2-Chloro-3-fluoro-4-(trimethylsilyl)pyridine ≥97%
2-Chloro-3-fluoro-4-(trimethylsilyl)pyridine

Precaución: For Research Use Only

Formula: 203.717 g/mol
MW: C₈H₁₁ClFNSi
Storage Temperature: Ambient
MDL Number: MFCD11043672
CAS Number: 860296-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR