1-(3,4-Dimethylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139912-5G 139912-1G 139912-25G 139912-500MG
77631-624EA 904 USD
77631-624 77636-602 77657-430 77648-612
1-(3,4-Dimethylphenyl)-2-methylpropan-1-ol ≥95%
1-(3,4-Dimethylphenyl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: 178.271 g/mol
MW: C₁₂H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD11934821
CAS Number: 1181615-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR