2-Methyl-1-(4-phenoxyphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139895-5G 139895-1G 139895-25G 139895-500MG
77631-270EA 1658.3 USD
77631-270 77636-600 77653-516 77648-608
2-Methyl-1-(4-phenoxyphenyl)propan-1-ol ≥95%
2-Methyl-1-(4-phenoxyphenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 242.313 g/mol
MW: C₁₆H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD13474847
CAS Number: 1282136-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR